Assay Details
| Gene Symbol: ARSG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048098.1 | NP_001041563.1 | 985 | 62 | ||
| XM_005624176.3 | XP_005624233.1 | 1553 | 62 | |||
| XM_022422567.1 | XP_022278275.1 | 1558 | 62 | |||
More Information
Set Membership: |
|

ARSG
Genomic Map