Assay Details
| Gene Symbol: ARSH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048123.1 | NP_001041588.1 | 2 - 3 | 212 | 72 | |
| XM_005641000.3 | XP_005641057.1 | 4 - 5 | 433 | 72 | ||
| XM_005641001.3 | XP_005641058.1 | 4 - 5 | 410 | 72 | ||
More Information
Set Membership: |
|

ARSH
Genomic Map