Assay Details
| Gene Symbol: ARSH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048123.1 | NP_001041588.1 | 3 - 4 | 341 | 89 | |
| XM_005641000.3 | XP_005641057.1 | 5 - 6 | 562 | 89 | ||
| XM_005641001.3 | XP_005641058.1 | 5 - 6 | 539 | 89 | ||
More Information
Set Membership: |
|

ARSH
Genomic Map