Assay Details
| Gene Symbol: ARSH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048123.1 | NP_001041588.1 | 7 - 8 | 1200 | 118 | |
| XM_005641000.3 | XP_005641057.1 | 9 - 10 | 1421 | 118 | ||
| XM_005641001.3 | XP_005641058.1 | 9 - 10 | 1398 | 118 | ||
More Information
Set Membership: |
|

ARSH
Genomic Map