Assay Details
| Gene Symbol: ARSH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048123.1 | NP_001041588.1 | 8 - 9 | 1325 | 55 | |
| XM_005641000.3 | XP_005641057.1 | 10 - 11 | 1546 | 55 | ||
| XM_005641001.3 | XP_005641058.1 | 10 - 11 | 1523 | 55 | ||
More Information
Set Membership: |
|

ARSH
Genomic Map