Assay Details
| Gene Symbol: ANGPT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001048126.1 | NP_001041591.1 | 6 - 7 | 1049 | 73 | |
| XM_005629929.3 | XP_005629986.1 | 5 - 6 | 1527 | 73 | ||
| XM_022403615.1 | XP_022259323.1 | 5 - 6 | 729 | 73 | ||
| XM_022403616.1 | XP_022259324.1 | 5 - 6 | 1017 | 73 | ||
| GenBank mRNA | AB256019.1 | - | 6 - 7 | 1049 | 73 | |
More Information
Set Membership: |
|

ANGPT2
Genomic Map