Assay Details
| Gene Symbol: SERPINA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080109.2 | NP_001073578.2 | 2 - 3 | 691 | 72 | |
| XM_022421757.1 | XP_022277465.1 | 2 - 3 | 781 | 72 | ||
| GenBank mRNA | AB287507.1 | - | 1 - 2 | 643 | 72 | |
More Information
Set Membership: |
|

SERPINA1
Genomic Map