Assay Details
| Gene Symbol: B3GALT4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080722.1 | NP_001074191.1 | 1 | 1344 | 61 | |
| XM_005627070.3 | XP_005627127.1 | 2 | 1256 | 61 | ||
| XM_014118044.2 | XP_013973519.1 | 2 | 1286 | 61 | ||
More Information
Set Membership: |
|

B3GALT4
Genomic Map