Assay Details
| Gene Symbol: CRYGC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001082600.1 | NP_001076069.1 | 2 - 3 | 341 | 73 | |
| Gene Symbol: CRYGD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | EF426309.1 | - | 2 - 3 | 341 | 73 | |
More Information
Set Membership: |
|

CRYGC
Genomic Map