Assay Details
| Gene Symbol: PRCD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001097560.2 | NP_001091029.1 | 249 | 75 | ||
| XM_022422503.1 | XP_022278211.1 | 2130 | 75 | |||
| GenBank mRNA | DQ390330.1 | - | 249 | 75 | ||
| DQ390331.1 | - | 593 | 75 | |||
| DQ390332.1 | - | 113 | 75 | |||
| DQ390333.1 | - | 42 | 75 | |||
| DQ390334.1 | - | 80 | 75 | |||
| DQ390335.1 | - | 80 | 75 | |||
| DQ390336.1 | - | 80 | 75 | |||
More Information
Set Membership: |
|

PRCD
Genomic Map