Assay Details
| Gene Symbol: ANGPTL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001109813.1 | NP_001103283.1 | 4 - 5 | 1531 | 85 | |
| XM_005622263.3 | XP_005622320.1 | 3 - 4 | 1434 | 85 | ||
More Information
Set Membership: |
|

ANGPTL1
Genomic Map