Assay Details
| Gene Symbol: RPL27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003102.2 | NP_001003102.2 | 1 | 270 | 154 | |
| GenBank mRNA | AJ388516.1 | - | 1 | 249 | 154 | |
| DN875070.1 | - | 1 | 247 | 154 | ||
More Information
Set Membership: |
|

RPL27
Genomic Map