Assay Details
| Gene Symbol: FGF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003432481.4 | XP_003432529.3 | 1 | 392 | 147 | |
| GenBank mRNA | AF060562.1 | - | 1 | 204 | 147 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: FGF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003432481.4 | XP_003432529.3 | 1 | 392 | 147 | |
| GenBank mRNA | AF060562.1 | - | 1 | 204 | 147 | |
Set Membership: |
|