Assay Details
| Gene Symbol: CABYR | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001145070.1 | NP_001138542.1 | 205 | 95 | ||
| XM_005622988.3 | XP_005623045.1 | 365 | 95 | |||
| XM_005622989.3 | XP_005623046.1 | 383 | 95 | |||
| XM_005622991.3 | XP_005623048.1 | 365 | 95 | |||
| XM_005622992.3 | XP_005623049.1 | 365 | 95 | |||
| XM_014115497.2 | XP_013970972.1 | 167 | 95 | |||
| XM_022420712.1 | XP_022276420.1 | 274 | 95 | |||
| XM_022420713.1 | XP_022276421.1 | 460 | 95 | |||
| GenBank mRNA | AM260639.1 | - | 205 | 95 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_014115497.2 transcripts.
Set Membership: |
|

CABYR
Genomic Map