Assay Details
| Gene Symbol: GREM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005622573.2 | XP_005622630.1 | 2 | 391 | 88 | |
| XM_022421119.1 | XP_022276827.1 | 3 | 561 | 88 | ||
More Information
Set Membership: |
|

GREM2
Genomic Map