Assay Details
| Gene Symbol: LOC403581 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_036671.1 | 1 - 2 | 106 | 98 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | alpha-1,3-mannosyl-glycoprotein 4-beta-N-acetylglucosaminyltransferase-like protein MGAT4E | LOC102151492 | -65.6kb | Sense | Chr. 7 - 321551 - 327610 on NCBI Build CanFam3.1 | |
| Downstream | synaptotagmin 2 | SYT2 | 103.5kb | Anti-sense | Chr. 7 - 497387 - 509338 on NCBI Build CanFam3.1 |
More Information
Set Membership: |
|

LOC403581
Genomic Map