Assay Details
| Gene Symbol: TNFSF13B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161710.2 | NP_001155182.1 | 5 - 6 | 995 | 94 | |
| XM_022408006.1 | XP_022263714.1 | 4 - 5 | 1334 | 94 | ||
| GenBank mRNA | FJ793193.2 | - | 5 - 6 | 995 | 94 | |
More Information
Set Membership: |
|

TNFSF13B
Genomic Map