Assay Details
| Gene Symbol: EIF2AK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161713.1 | NP_001155185.1 | 1 | 284 | 60 | |
| GenBank mRNA | FJ911905.1 | - | 1 | 284 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EIF2AK1| Gene Symbol: EIF2AK1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001161713.1 | NP_001155185.1 | 1 | 284 | 60 | |
| GenBank mRNA | FJ911905.1 | - | 1 | 284 | 60 | |
Set Membership: |
|