Assay Details
| Gene Symbol: GPX3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001164454.1 | NP_001157926.1 | 2 - 3 | 293 | 69 | |
| GenBank mRNA | DN374277.1 | - | 2 - 3 | 291 | 69 | |
| DN439024.1 | - | 2 - 3 | 303 | 69 | ||
| DN904267.1 | - | 2 - 3 | 286 | 69 | ||
More Information
Set Membership: |
|

GPX3
Genomic Map