Assay Details
| Gene Symbol: IL17A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001165878.1 | NP_001159350.1 | 2 - 3 | 246 | 75 | |
| GenBank mRNA | AB514445.1 | - | 2 - 3 | 246 | 75 | |
More Information
Set Membership: |
|

IL17A
Genomic Map