Assay Details
| Gene Symbol: SPAG11B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171544.1 | NP_001165015.1 | 2 - 3 | 203 | 104 | |
| GenBank mRNA | DQ012011.1 | - | 2 - 3 | 203 | 104 | |
More Information
Set Membership: |
|

SPAG11B
Genomic Map