Assay Details
| Gene Symbol: SPAG11E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171545.1 | NP_001165016.1 | 1 - 2 | 55 | 84 | |
| GenBank mRNA | DQ012012.1 | - | 1 - 2 | 55 | 84 | |
More Information
Set Membership: |
|

SPAG11E
Genomic Map