Assay Details
| Gene Symbol: RBM12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001171748.1 | NP_001165219.1 | 3 | 2852 | 105 | |
| GenBank mRNA | DQ341269.1 | - | 3 | 2852 | 105 | |
More Information
Set Membership: |
|

RBM12
Genomic Map