Assay Details
| Gene Symbol: KLK8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197162.1 | NP_001184091.1 | 2 - 3 | 237 | 91 | |
| NM_001197163.1 | NP_001184092.1 | 2 - 3 | 237 | 91 | ||
| XM_005615762.2 | XP_005615819.1 | 3 - 4 | 374 | 91 | ||
| XM_014114292.2 | XP_013969767.1 | 3 - 4 | 360 | 91 | ||
| GenBank mRNA | HM135456.1 | - | 2 - 3 | 237 | 91 | |
| HM135457.1 | - | 2 - 3 | 237 | 91 | ||
More Information
Set Membership: |
|

KLK8
Genomic Map