Assay Details
| Gene Symbol: KLK8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197162.1 | NP_001184091.1 | 4 - 5 | 622 | 74 | |
| XM_005615762.2 | XP_005615819.1 | 5 - 6 | 759 | 74 | ||
| XM_014114292.2 | XP_013969767.1 | 5 - 6 | 745 | 74 | ||
| GenBank mRNA | HM135456.1 | - | 4 - 5 | 622 | 74 | |
More Information
Set Membership: |
|

KLK8
Genomic Map