Assay Details
| Gene Symbol: TLR5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001197176.1 | NP_001184105.1 | 1 | 2153 | 100 | |
| XM_005640757.2 | XP_005640814.1 | 2 | 2299 | 100 | ||
| XM_022415290.1 | XP_022270998.1 | 2 | 4471 | 100 | ||
More Information
Set Membership: |
|

TLR5
Genomic Map