Assay Details
| Gene Symbol: FPGT | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001204810.1 | NP_001191739.1 | 2 - 3 | 250 | 90 | |
| XM_022419470.1 | XP_022275178.1 | 2 - 3 | 306 | 90 | ||
More Information
Set Membership: |
|

FPGT
Genomic Map