Assay Details
| Gene Symbol: TNFSF12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205168.1 | NP_001192097.1 | 6 - 7 | 496 | 57 | |
| XM_005619471.3 | XP_005619528.1 | 4 - 5 | 459 | 57 | ||
| GenBank mRNA | FJ915118.1 | - | 6 - 7 | 496 | 57 | |
More Information
Set Membership: |
|

TNFSF12
Genomic Map