Assay Details
| Gene Symbol: MEF2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205203.1 | NP_001192132.1 | 2 - 3 | 260 | 68 | |
| XM_022406260.1 | XP_022261968.1 | 3 - 4 | 1166 | 68 | ||
| XM_022406261.1 | XP_022261969.1 | 3 - 4 | 429 | 68 | ||
More Information
Set Membership: |
|

MEF2B
Genomic Map