Assay Details
| Gene Symbol: GGTA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206937.1 | NP_001193866.1 | 1 - 2 | 114 | 108 | |
| XM_005625025.3 | XP_005625082.1 | 2 - 3 | 191 | 108 | ||
| XM_005625027.2 | XP_005625084.1 | 3 - 4 | 384 | 108 | ||
| XM_005625028.3 | XP_005625085.1 | 2 - 3 | 290 | 108 | ||
| XM_005625029.3 | XP_005625086.1 | 2 - 3 | 223 | 108 | ||
| XM_005625030.3 | XP_005625087.1 | 2 - 3 | 244 | 108 | ||
| XM_005625032.3 | XP_005625089.1 | 3 - 4 | 243 | 108 | ||
| XM_014116859.2 | XP_013972334.1 | 2 - 3 | 227 | 108 | ||
| XM_014116860.2 | XP_013972335.1 | 2 - 3 | 227 | 108 | ||
| XM_022422600.1 | XP_022278308.1 | 3 - 4 | 321 | 108 | ||
| XM_022422602.1 | XP_022278310.1 | 3 - 4 | 319 | 108 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001206937.1, XM_005625025.3, XM_005625027.2, XM_005625028.3, XM_005625029.3, XM_005625030.3, XM_005625032.3, XM_014116859.2, XM_014116860.2, XM_022422600.1, XM_022422602.1 transcripts.
Set Membership: |
|

GGTA1
Genomic Map