Assay Details
| Gene Symbol: GGTA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206937.1 | NP_001193866.1 | 5 - 6 | 410 | 65 | |
| XM_005625025.3 | XP_005625082.1 | 7 - 8 | 541 | 65 | ||
| XM_005625027.2 | XP_005625084.1 | 8 - 9 | 734 | 65 | ||
| XM_005625028.3 | XP_005625085.1 | 7 - 8 | 640 | 65 | ||
| XM_005625029.3 | XP_005625086.1 | 7 - 8 | 573 | 65 | ||
| XM_005625030.3 | XP_005625087.1 | 7 - 8 | 594 | 65 | ||
| XM_005625032.3 | XP_005625089.1 | 8 - 9 | 593 | 65 | ||
| XM_014116859.2 | XP_013972334.1 | 6 - 7 | 523 | 65 | ||
| XM_014116860.2 | XP_013972335.1 | 5 - 6 | 487 | 65 | ||
| XM_014116862.2 | XP_013972337.1 | 7 - 8 | 426 | 65 | ||
| XM_014116863.2 | XP_013972338.1 | 5 - 6 | 350 | 65 | ||
| XM_022422600.1 | XP_022278308.1 | 8 - 9 | 671 | 65 | ||
| XM_022422601.1 | XP_022278309.1 | 6 - 7 | 369 | 65 | ||
| XM_022422602.1 | XP_022278310.1 | 7 - 8 | 615 | 65 | ||
More Information
Set Membership: |
|

GGTA1
Genomic Map