Assay Details
| Gene Symbol: GGTA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206937.1 | NP_001193866.1 | 6 - 7 | 542 | 112 | |
| XM_005625025.3 | XP_005625082.1 | 8 - 9 | 673 | 112 | ||
| XM_005625027.2 | XP_005625084.1 | 9 - 10 | 866 | 112 | ||
| XM_005625028.3 | XP_005625085.1 | 8 - 9 | 772 | 112 | ||
| XM_005625029.3 | XP_005625086.1 | 8 - 9 | 705 | 112 | ||
| XM_005625030.3 | XP_005625087.1 | 8 - 9 | 726 | 112 | ||
| XM_005625032.3 | XP_005625089.1 | 9 - 10 | 725 | 112 | ||
| XM_014116859.2 | XP_013972334.1 | 7 - 8 | 655 | 112 | ||
| XM_014116860.2 | XP_013972335.1 | 6 - 7 | 619 | 112 | ||
| XM_014116862.2 | XP_013972337.1 | 8 - 9 | 558 | 112 | ||
| XM_014116863.2 | XP_013972338.1 | 6 - 7 | 482 | 112 | ||
| XM_022422600.1 | XP_022278308.1 | 9 - 10 | 803 | 112 | ||
| XM_022422601.1 | XP_022278309.1 | 7 - 8 | 501 | 112 | ||
| XM_022422602.1 | XP_022278310.1 | 8 - 9 | 747 | 112 | ||
More Information
Set Membership: |
|

GGTA1
Genomic Map