Assay Details
| Gene Symbol: SELPLG | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001242719.1 | NP_001229648.1 | 1 | 1126 | 103 | |
| XM_005636028.3 | XP_005636085.1 | 2 | 1305 | 103 | ||
| GenBank mRNA | AB614446.1 | - | 1 | 1126 | 103 | |
More Information
Set Membership: |
|

SELPLG
Genomic Map