Assay Details
| Gene Symbol: VDAC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251965.2 | NP_001238894.1 | 1 | 627 | 78 | |
| NM_001251966.2 | NP_001238895.1 | 1 | 624 | 78 | ||
| GenBank mRNA | DN408490.1 | - | 1 | 309 | 78 | |
More Information
Set Membership: |
|

VDAC3
Genomic Map