Assay Details
| Gene Symbol: MYL9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252166.1 | NP_001239095.1 | 2 - 3 | 293 | 87 | |
| XM_014106841.2 | XP_013962316.1 | 2 - 3 | 596 | 87 | ||
| GenBank mRNA | DN399972.1 | - | 2 - 3 | 293 | 87 | |
More Information
Set Membership: |
|

MYL9
Genomic Map