Assay Details
| Gene Symbol: MYL9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252166.1 | NP_001239095.1 | 3 - 4 | 458 | 65 | |
| XM_014106841.2 | XP_013962316.1 | 3 - 4 | 761 | 65 | ||
| GenBank mRNA | CX003111.1 | - | 1 - 2 | 694 | 65 | |
| DN399972.1 | - | 3 - 4 | 458 | 65 | ||
More Information
Set Membership: |
|

MYL9
Genomic Map