Assay Details
| Gene Symbol: FYTTD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251944.1 | NP_001238873.1 | 1 | 1018 | 114 | |
| GenBank mRNA | DN350103.1 | - | 1 | 417 | 114 | |
More Information
Set Membership: |
|

FYTTD1
Genomic Map