Assay Details
| Gene Symbol: OTUB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251947.1 | NP_001238876.1 | 7 | 921 | 115 | |
| GenBank mRNA | CO595103.1 | - | 1 | 75 | 115 | |
| Gene Symbol: OTUB1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_022405754.1 | XP_022261462.1 | 7 | 934 | 115 | |
| XM_022405755.1 | XP_022261463.1 | 7 | 1153 | 115 | ||
More Information
Set Membership: |
|

OTUB1
Genomic Map