Assay Details
| Gene Symbol: GSTP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252167.1 | NP_001239096.1 | 7 | 720 | 102 | |
| XM_005631040.2 | XP_005631097.1 | 7 | 757 | 102 | ||
| GenBank mRNA | CO700565.1 | - | 1 | 83 | 102 | |
More Information
Set Membership: |
|

GSTP1
Genomic Map