Assay Details
| Gene Symbol: NPC2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001003242.2 | NP_001003242.2 | 2 - 3 | 259 | 67 | |
| GenBank mRNA | DN349868.1 | - | 2 - 3 | 259 | 67 | |
| DN392295.1 | - | 2 - 3 | 210 | 67 | ||
| S77411.1 | - | 2 - 3 | 209 | 67 | ||
More Information
Set Membership: |
|

NPC2
Genomic Map