Assay Details
| Gene Symbol: RPL18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001251958.1 | NP_001238887.1 | 5 - 6 | 447 | 69 | |
| GenBank mRNA | DN371043.1 | - | 5 - 6 | 447 | 69 | |
| DQ403035.1 | - | 4 - 5 | 354 | 69 | ||
More Information
Set Membership: |
|

RPL18
Genomic Map