Assay Details
| Gene Symbol: FKBP1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252190.3 | NP_001239119.1 | 1 | 112 | 77 | |
| GenBank mRNA | DN392962.1 | - | 2 | 150 | 77 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.
| Gene Symbol: FKBP1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001252190.3 | NP_001239119.1 | 1 | 112 | 77 | |
| GenBank mRNA | DN392962.1 | - | 2 | 150 | 77 | |
Set Membership: |
|