Assay Details
| Gene Symbol: KRT14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001253741.1 | NP_001240670.1 | 1410 | 84 | ||
| GenBank mRNA | DN872732.1 | - | 643 | 84 | ||
| DN873542.1 | - | 273 | 84 | |||
More Information
Set Membership: |
|

KRT14
Genomic Map