Assay Details
| Gene Symbol: RFX8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001270887.1 | NP_001257816.1 | 7 - 8 | 775 | 60 | |
| XM_022423975.1 | XP_022279683.1 | 10 - 11 | 1012 | 60 | ||
More Information
Set Membership: |
|

RFX8
Genomic Map