Assay Details
Gene Symbol: UCN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005632587.3 | XP_005632644.1 | 1 | 714 | 102 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: UCN2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005632587.3 | XP_005632644.1 | 1 | 714 | 102 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |