Assay Details
| Gene Symbol: LTA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001313849.1 | NP_001300778.1 | 2 - 3 | 203 | 65 | |
| XM_843793.4 | XP_848886.2 | 3 - 4 | 654 | 65 | ||
More Information
Set Membership: |
|

LTA
Genomic Map