Assay Details
| Gene Symbol: CCR2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_005632630.3 | XP_005632687.1 | 1 - 2 | 86 | 67 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005632630.3 transcripts.
Set Membership: |
|

CCR2
Genomic Map