Assay Details
| Gene Symbol: MIR187 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_049431.1 | 1 | 36 | 57 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | ribosome biogenesis protein NSA2 homolog | LOC100683727 | -3.5mb | Anti-sense | Chr. 7 - 48880428 - 48896436 on NCBI Build CanFam3.1 | |
| Downstream | uncharacterized LOC106558998 | LOC106558998 | 357.6kb | Sense | Chr. 7 - 52715568 - 52716484 on NCBI Build CanFam3.1 |
More Information
Set Membership: |
|

MIR187
Genomic Map