Assay Details
| Gene Symbol: MIR302D | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_049446.1 | 1 | 31 | 69 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | microRNA mir-367 | MIR367 | cfa-mir-367 | Sense | Chr. 32 - 32428905 - 32428972 on NCBI Build CanFam3.1 |
More Information
Set Membership: |
|

MIR302D
Genomic Map