Assay Details
| Gene Symbol: MIR208A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_049457.1 | 1 | 34 | 64 | ||
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | cleavage and polyadenylation specific factor 2 | CPSF2 | -78.3kb | Anti-sense | Chr. 8 - 1364399 - 1398496 on NCBI Build CanFam3.1 | |
| Downstream | solute carrier family 24 member 4 | SLC24A4 | 93.9kb | Anti-sense | Chr. 8 - 1570833 - 1685135 on NCBI Build CanFam3.1 |
More Information
Set Membership: |
|

MIR208A
Genomic Map